missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride

Catalog No. p-204137
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175110250 25 mL
AC175111000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175110250 Supplier Thermo Scientific Chemicals Supplier No. 175110250
Only null left

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Specifications

Chemical Name or Material Triethyloxonium tetrafluoroborate
Name Note 1M Solution in Methylene Chloride
CAS 368-39-8, 75-09-2
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3OBF4
MDL Number MFCD00044423
Fieser 01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337
Density 1.3280g/mL
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
Solubility Information Solubility in water: reacts.
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CC[O+](CC)CC
IUPAC Name tetrafluoroboranuide; triethyloxidanium
Molecular Weight (g/mol) 189.99
PubChem CID 2723982
Formula Weight 189.99
Packaging Plastic bottle
Specific Gravity 1.328
Quantity 25 mL
Show More Show Less

RUO – Research Use Only