missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic Acid 98.0+%, TCI America™
Supplier: TCI America T08065G
Specifications
| Triethylenetetramine-N,N,N′,N′′,N′′′,N′′′-hexa | |
| White | |
| MFCD00004290 | |
| ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid | |
| C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O | |
| 494.454 | |
| 494.45 | |
| Crystalline Powder |
| 869-52-3 | |
| C18H30N4O12 | |
| 5 g | |
| RAEOEMDZDMCHJA-UHFFFAOYSA-N | |
| 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid | |
| 70088 | |
| ≥98.0% (T) |
Chemical Identifiers
| 869-52-3 | |
| 494.454 | |
| RAEOEMDZDMCHJA-UHFFFAOYSA-N | |
| 70088 | |
| C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O |
| C18H30N4O12 | |
| MFCD00004290 | |
| ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid | |
| 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid |
Safety and Handling
TSCA : Yes