missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic Acid 98.0+%, TCI America™

Catalog No. p-7136439
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08065G 5 g
T080625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08065G Supplier TCI America Supplier No. T08065G
Only null left

Chemical Identifiers

CAS 869-52-3
Molecular Formula C18H30N4O12
Molecular Weight (g/mol) 494.454
MDL Number MFCD00004290
InChI Key RAEOEMDZDMCHJA-UHFFFAOYSA-N
Synonym ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid
PubChem CID 70088
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 494.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylenetetramine-N,N,N′,N′′,N′′′,N′′′-hexa