missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Monomethyl Ether 98.0+%, TCI America™

Catalog No. p-7136401
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T070925ML 25 mL
T0709500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T070925ML Supplier TCI America Supplier No. T070925ML
Only null left

Chemical Identifiers

CAS 112-35-6
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.201
MDL Number MFCD00002875
InChI Key JLGLQAWTXXGVEM-UHFFFAOYSA-N
Synonym Methyl Triglycol, mPEG3-Alcohol
PubChem CID 8178
ChEBI CHEBI:84233
IUPAC Name 2-[2-(2-methoxyethoxy)ethoxy]ethanol
SMILES COCCOCCOCCO

Specifications

Melting Point -45°C
Color Colorless
Boiling Point 248°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monomethyl Ether
EINECSNumber (2)-0442&(7)-0097
RTECSNumber KL6390000
TSCA Yes