accessibility menu, dialog, popup

UserName

CAS RN 112-35-6

H3COOOOH

CAS RN 112-35-6

IUPAC Name: 2-[2-(2-methoxyethoxy)ethoxy]ethan-1-ol
Synonyms: methoxy triethylene glycol
Molecular Weight (g/mol): 164.2
Molecular Formula: C7H16O4
InChi Key: JLGLQAWTXXGVEM-UHFFFAOYSA-N
SMILES: COCCOCCOCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results