missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Monobenzyl Ether 97.0+%, TCI America™
$315.03 - $1252.65
Chemical Identifiers
| CAS | 55489-58-2 |
|---|---|
| Molecular Formula | C13H20O4 |
| Molecular Weight (g/mol) | 240.299 |
| MDL Number | MFCD02093556 |
| InChI Key | KVPHZILZNXDCNH-UHFFFAOYSA-N |
| Synonym | 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol |
| PubChem CID | 2737322 |
| IUPAC Name | 2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol |
| SMILES | C1=CC=C(C=C1)COCCOCCOCCO |
Chemical Identifiers
| 55489-58-2 | |
| 240.299 | |
| KVPHZILZNXDCNH-UHFFFAOYSA-N | |
| 2737322 | |
| C1=CC=C(C=C1)COCCOCCOCCO |
| C13H20O4 | |
| MFCD02093556 | |
| 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol | |
| 2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol |
Specifications
| 55489-58-2 | |
| C13H20O4 | |
| 5 g | |
| KVPHZILZNXDCNH-UHFFFAOYSA-N | |
| 2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol | |
| 2737322 | |
| ≥97.0% (GC) | |
| Triethylene Glycol Monobenzyl Ether |
| Yellow | |
| MFCD02093556 | |
| 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol | |
| C1=CC=C(C=C1)COCCOCCOCCO | |
| 240.299 | |
| 240.30 | |
| Liquid |
Safety and Handling
TSCA : No