missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Mono(2-propynyl) Ether 96.0+%, TCI America™
$169.63 - $529.40
Chemical Identifiers
| CAS | 208827-90-1 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.223 |
| MDL Number | MFCD13189984 |
| InChI Key | CXJWUJYYDLYCCQ-UHFFFAOYSA-N |
| Synonym | 2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl Ether |
| PubChem CID | 11506589 |
| IUPAC Name | 2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethanol |
| SMILES | C#CCOCCOCCOCCO |
Chemical Identifiers
| 208827-90-1 | |
| 188.223 | |
| CXJWUJYYDLYCCQ-UHFFFAOYSA-N | |
| 11506589 | |
| C#CCOCCOCCOCCO |
| C9H16O4 | |
| MFCD13189984 | |
| 2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl Ether | |
| 2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethanol |
Specifications
| 208827-90-1 | |
| C9H16O4 | |
| 1 g | |
| CXJWUJYYDLYCCQ-UHFFFAOYSA-N | |
| 2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethanol | |
| 11506589 | |
| ≥96.0% (GC) | |
| Triethylene Glycol Mono(2-propynyl) Ether |
| Yellow | |
| MFCD13189984 | |
| 2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl Ether | |
| C#CCOCCOCCOCCO | |
| 188.223 | |
| 188.22 | |
| Liquid |
Safety and Handling
TSCA : No