accessibility menu, dialog, popup

UserName

HCOOOOH

CAS RN 208827-90-1

IUPAC Name: 2-{2-[2-(prop-2-yn-1-yloxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[2-(prop-2-yn-1-yloxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: CXJWUJYYDLYCCQ-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results