missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethyl Ether (stabilized with BHT) 99.0+%, TCI America™

Catalog No. B049625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B049625G 25 g
B0496500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B049625G Supplier TCI America Supplier No. B049625G

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.228
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Melting Point -45°C
Color Colorless
Boiling Point 216°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Dimethyl Ether (stabilized with BHT)
EINECSNumber (7)-1321
RTECSNumber XF0665000
TSCA Yes