missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethyl ether, 99%

CAS: 112-49-2 | C8H18O4 | 178.228 g/mol

$70.78 - $464.27

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.228
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0412822
View Documents
Thermo Scientific Chemicals
L0412822
100 g
Each for $70.78
Only null left
 
AAL0412836
View Documents
Thermo Scientific Chemicals
L0412836
500 g
Each for $156.40
Only null left
 
AAL041280E
View Documents
Thermo Scientific Chemicals
L041280E
2500 g
Each for $464.27
Only null left
 
Description

Description

Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

Solubility
Miscible with water, chloroform and methanol

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

112-49-2
178.228
YFNKIDBQEZZDLK-UHFFFAOYSA-N
8189
1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
C8H18O4
MFCD00008504
triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
CHEBI:44842
COCCOCCOCCOC

Specifications

112-49-2
0.986
113°C (235°F)
1.423
MFCD00008504
1700630
14,9692
Miscible with water,chloroform and methanol
COCCOCCOCCOC
178.228
CHEBI:44842
99%
-45°C
215°C to 216°C
C8H18O4
CH3O(CH2CH2O)3CH3
100 g
Hygroscopic
triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
YFNKIDBQEZZDLK-UHFFFAOYSA-N
1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
8189
178.23
Triethylene glycol dimethyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H360Df

EINECSNumber : 203-977-3

RTECSNumber : XF0665000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only