missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Bis(p-toluenesulfonate) 97.0+%, TCI America™

Catalog No. p-7136801
Change view
Click to view available options
Quantity:
10 g
25 g
Catalog No. Quantity
T148810G 10 g
T148825G 25 g
2 options

Catalog No. T148810G

Supplier: TCI America T148810G

Only null left

Chemical Identifiers

CAS 19249-03-7
Molecular Formula C20H26O8S2
Molecular Weight (g/mol) 458.54
MDL Number MFCD00048096
InChI Key KCONMNWPRXAWKK-UHFFFAOYSA-N
Synonym tri ethylene glycol di-p-toluenesulfonate, triethylene glycol ditosylate, ethane-1,2-diylbis oxy bis ethane-2,1-diyl bis 4-methylbenzenesulfonate, triethylene glycol di p-toluenesulfonate, 1,2-bis 2-tosyloxyethoxy ethane, 2-2-2-4-methylbenzenesulfonyl oxy ethoxy ethoxy ethyl 4-methylbenzene-1-sulfonate, ethane-1,2-diylbis oxyethane-2,1-diyl bis 4-methylbenzenesulfonate, 2-2-2-4-methylphenyl sulfonyloxy ethoxy ethoxy ethyl 4-methylbenzenesulfon ate, 2-2-2-4-methylphenyl sulfonyloxyethoxy ethoxy ethyl 4-methylbenzenesulfonate, 2-2-2-4-methylbenzenesulfonyl oxy ethoxy ethoxy ethyl 4-methylbenzenesulfonate
PubChem CID 316652
IUPAC Name 2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 82°C
Color White
Quantity 10 g
Formula Weight 458.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triethylene Glycol Bis(p-toluenesulfonate)