accessibility menu, dialog, popup

UserName

H3CSOOOOOOSOOCH3

CAS RN 19249-03-7

IUPAC Name: 2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 458.54
Molecular Formula: C20H26O8S2
InChi Key: KCONMNWPRXAWKK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)