missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Triethylamine Acetate (TEAA) 2.0 M

Catalog No. 400613
Encompass
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400613 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 400613 Supplier Applied Biosystems™ Supplier No. 400613

This 2.0 M Triethylamine Acetate solution is used with OPC™ cartridges (Cat. No. 400771), for oligonucleotide purification.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Specifications

Chemical Name or Material Triethylamine Acetate
Content And Storage Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.
Format Bottle
Sample Type DNA
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
Product Type Oligonucleotide Synthesis Reagent
Quantity 200 mL
Reagent Type Solvent or General Reagent
Shipping Condition Room Temperature
Form Liquid
Show More Show Less
Do you know of any alternatives for Triethylamine Acetate (TEAA) 2.0 M (Cat. No. 400613)?

We do not offer any alternatives for Triethylamine Acetate (TEAA) 2.0 M.

For Research Use Only. Not for use in diagnostic procedures.