missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylamine, ≥99.5% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 734875ML
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| 593°F | |
| 51.75 mmHg (20°C) | |
| 0.726 g/mL (at 25°C) | |
| Neat | |
| MFCD00009051 | |
| 88.8°C | |
| Analytical Standard | |
| 101.19 | |
| Analytical Standard |
| 121-44-8 | |
| 3.5 (vs. air) | |
| -11°C | |
| (C2H5)3 N | |
| -115°C | |
| N,N-Diethylethanamine | |
| 5 mL | |
| ≥99.5% (GC) |