missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Phosphate 99.0+%, TCI America™

Catalog No. P027025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P027025G 25 g
P0270500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P027025G Supplier TCI America Supplier No. P027025G

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 216°C
Quantity 25 g
Formula Weight 182.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Phosphate
EINECSNumber (2)-2000
RTECSNumber TC7900000
TSCA Yes