missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphate, 99%

Catalog No. p-203956
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139680010 1 kg Glass bottle
AC139680025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139680010 Supplier Thermo Scientific Chemicals Supplier No. 139680010
Only null left

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Melting Point -56°C
Density 1.0700g/mL
Boiling Point 215°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.407
Linear Formula (C2H5O)3P(O)
Quantity 1 kg
Beilstein 01, 332
Fieser 01,1212; 16,3
Specific Gravity 1.07
Merck Index 15, 9838
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols,ethers,ketones,esters,,aliphatic,aromatic and chlorinated hydrocarbons,soluble in most other organic solvents
Formula Weight 182.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethyl phosphate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-114-5
RTECSNumber TC7900000
TSCA TSCA

RUO – Research Use Only