missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthopropionate 96.0+%, TCI America™

Catalog No. O006725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O006725ML 25 mL
O0067500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O006725ML Supplier TCI America Supplier No. O006725ML

Chemical Identifiers

CAS 115-80-0
Molecular Formula C9H20O3
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009226
InChI Key FGWYWKIOMUZSQF-UHFFFAOYSA-N
Synonym triethyl orthopropionate, ethyl orthopropionate, propane, 1,1,1-triethoxy, triethylorthopropionate, triethyl o-propionate, orthopropionic acid ethyl ester, orthopropionic acid, triethyl ester, unii-9uvm48bas9, 9uvm48bas9, 1,1,1-triethoxypropan
PubChem CID 66995
IUPAC Name 1,1,1-triethoxypropane
SMILES CCOC(CC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 158°C
Quantity 25 mL
UN Number 3272
Formula Weight 176.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthopropionate
EINECSNumber (2)-0503
TSCA Yes