accessibility menu, dialog, popup

UserName

CH3OH3COH3COH3C

CAS RN 115-80-0

IUPAC Name: 1,1,1-triethoxypropane
Synonyms: triethylorthopropionate
Molecular Weight (g/mol): 176.26
Molecular Formula: C9H20O3
InChi Key: FGWYWKIOMUZSQF-UHFFFAOYSA-N
SMILES: CCOC(CC)(OCC)OCC
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)