missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthopropionate, 97%

Catalog No. AAL08172AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08172AE 100 mL
AAL08172AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08172AE Supplier Thermo Scientific Chemicals Supplier No. L08172AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115-80-0 | C9H20O3 | 176.26 g/mol

Used as intermediates in pharmaceutical chemical and organic synthesize. It can react with N-benzoyl-glycine in the presence of 4-(dimethylamino)pyridine and acetic anhydride to produce 4-(1-ethoxy-propylidene)-2-phenyl-4H-oxazol-5-one. It is also applied as a starting material in the synthesis of Triethyl orthoacrylate. Triethyl orthopropionate is also commonly used in Claisen rearrangement reactions that yield γ,δ-unsaturated esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediates in pharmaceutical chemical and organic synthesize. It can react with N-benzoyl-glycine in the presence of 4-(dimethylamino)pyridine and acetic anhydride to produce 4-(1-ethoxy-propylidene)-2-phenyl-4H-oxazol-5-one. It is also applied as a starting material in the synthesis of Triethyl orthoacrylate. Triethyl orthopropionate is also commonly used in Claisen rearrangement reactions that yield γ,δ-unsaturated esters.

Solubility
Hydrolyzes slowly in water. Soluble in alcohol, chloroform, ethyl acetate and ether.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect against electrostatic charges. Keep ignition sources away.

Chemical Identifiers

CAS 115-80-0
Molecular Formula C9H20O3
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009226
InChI Key FGWYWKIOMUZSQF-UHFFFAOYSA-N
Synonym triethyl orthopropionate, ethyl orthopropionate, propane, 1,1,1-triethoxy, triethylorthopropionate, triethyl o-propionate, orthopropionic acid ethyl ester, orthopropionic acid, triethyl ester, unii-9uvm48bas9, 9uvm48bas9, 1,1,1-triethoxypropan
PubChem CID 66995
IUPAC Name 1,1,1-triethoxypropane
SMILES CCOC(CC)(OCC)OCC

Specifications

Density 0.88
Boiling Point 158°C
Flash Point 45°C (113°F)
Odor Slightly Sweetish
Refractive Index 1.402
Linear Formula CH3CH2C(OCH2CH3)3
Quantity 100 mL
UN Number UN3272
Beilstein 906798
Solubility Information Hydrolyzes slowly in water. Soluble in alcohol,chloroform,ethyl acetate and ether.
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material Triethyl orthopropionate
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 204-108-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only