missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthobutyrate 90.0+%, TCI America™

Catalog No. p-7134223
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01735ML 5 mL
O017325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01735ML Supplier TCI America Supplier No. O01735ML

Chemical Identifiers

CAS 24964-76-9
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00059382
InChI Key KOPMZTKUZCNGFY-UHFFFAOYSA-N
Synonym Ethyl Orthobutyrate, Orthobutyric Acid Triethyl Ester, 1,1,1-Triethoxybutane
PubChem CID 371779
IUPAC Name 1,1,1-triethoxybutane
SMILES CCCC(OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 63°C
Quantity 5 mL
UN Number 3272
Formula Weight 190.28
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthobutyrate