accessibility menu, dialog, popup

UserName

CH3OH3COH3COH3C

CAS RN 24964-76-9

IUPAC Name: 1,1,1-triethoxybutane
Synonyms: 1,1,1-triethoxybutane
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: KOPMZTKUZCNGFY-UHFFFAOYSA-N
SMILES: CCCC(OCC)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results