missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl O-Acetylcitrate 97.0+%, TCI America™
$61.51 - $126.76
Chemical Identifiers
| CAS | 77-89-4 |
|---|---|
| Molecular Formula | C14H22O8 |
| Molecular Weight (g/mol) | 318.322 |
| MDL Number | MFCD00049378 |
| InChI Key | WEAPVABOECTMGR-UHFFFAOYSA-N |
| Synonym | acetyl triethyl citrate, triethyl o-acetylcitrate, citroflex a 2, acetyltriethyl citrate, atec, triethyl acetylcitrate, triethyl 2-acetoxypropane-1,2,3-tricarboxylate, o-acetylcitric acid triethyl ester, citric acid, acetyl triethyl ester, 1,2,3-propanetricarboxylic acid, 2-acetyloxy-, triethyl ester |
| PubChem CID | 6504 |
| IUPAC Name | triethyl 2-acetyloxypropane-1,2,3-tricarboxylate |
| SMILES | CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)OC(=O)C |
Chemical Identifiers
| 77-89-4 | |
| 318.322 | |
| WEAPVABOECTMGR-UHFFFAOYSA-N | |
| 6504 | |
| CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)OC(=O)C |
| C14H22O8 | |
| MFCD00049378 | |
| acetyl triethyl citrate, triethyl o-acetylcitrate, citroflex a 2, acetyltriethyl citrate, atec, triethyl acetylcitrate, triethyl 2-acetoxypropane-1,2,3-tricarboxylate, o-acetylcitric acid triethyl ester, citric acid, acetyl triethyl ester, 1,2,3-propanetricarboxylic acid, 2-acetyloxy-, triethyl ester | |
| triethyl 2-acetyloxypropane-1,2,3-tricarboxylate |
Specifications
| 77-89-4 | |
| 229°C | |
| MFCD00049378 | |
| acetyl triethyl citrate, triethyl o-acetylcitrate, citroflex a 2, acetyltriethyl citrate, atec, triethyl acetylcitrate, triethyl 2-acetoxypropane-1,2,3-tricarboxylate, o-acetylcitric acid triethyl ester, citric acid, acetyl triethyl ester, 1,2,3-propanetricarboxylic acid, 2-acetyloxy-, triethyl ester | |
| CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)OC(=O)C | |
| 318.322 | |
| 318.32 | |
| Liquid |
| Yellow | |
| C14H22O8 | |
| 25 g | |
| WEAPVABOECTMGR-UHFFFAOYSA-N | |
| triethyl 2-acetyloxypropane-1,2,3-tricarboxylate | |
| 6504 | |
| ≥97.0% (GC) | |
| Triethyl O-Acetylcitrate |
Safety and Handling
EINECSNumber : (2)-1327
RTECSNumber : GE8225000
TSCA : Yes