CAS RN 77-89-4
IUPAC Name:
1,2,3-triethyl 2-(acetyloxy)propane-1,2,3-tricarboxylate
Synonyms:
acetyl triethyl citrate
Molecular Weight (g/mol):
318.32
Molecular Formula:
C14H22O8
InChi Key:
WEAPVABOECTMGR-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(CC(=O)OCC)(OC(C)=O)C(=O)OCC
1
–
2
of
2
results
2
Triethyl O-Acetylcitrate 97.0+%, TCI America™
CAS: 77-89-4 Molecular Formula: C14H22O8 Molecular Weight (g/mol): 318.322 MDL Number: MFCD00049378 InChI Key: WEAPVABOECTMGR-UHFFFAOYSA-N Synonym: acetyl triethyl citrate,triethyl o-acetylcitrate,citroflex a 2,acetyltriethyl citrate,atec,triethyl acetylcitrate,triethyl 2-acetoxypropane-1,2,3-tricarboxylate,o-acetylcitric acid triethyl ester,citric acid, acetyl triethyl ester,1,2,3-propanetricarboxylic acid, 2-acetyloxy-, triethyl ester PubChem CID: 6504 IUPAC Name: triethyl 2-acetyloxypropane-1,2,3-tricarboxylate SMILES: CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)OC(=O)C