Learn More
Triethyl citrate, 99%
CAS: 77-93-0 | C12H20O7 | 276.285 g/mol
Supplier: Thermo Scientific Chemicals L126390B
Description
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.
Solubility
Miscible with water, alcohol and diethyl ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Triethyl citrate | |
| -55°C | |
| 126°C to 127°C (1 mmHg) | |
| C12H20O7 | |
| MFCD00009201 | |
| 1801199 | |
| triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
| DOOTYTYQINUNNV-UHFFFAOYSA-N | |
| triethyl 2-hydroxypropane-1,2,3-tricarboxylate | |
| 6506 | |
| 99% |
| 77-93-0 | |
| 1.137 | |
| 176°C (348°F) | |
| 1.442 | |
| 1000 g | |
| 14,2326 | |
| Miscible with water,alcohol and diethyl ether. | |
| CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O | |
| 276.285 | |
| 276.29 |
Chemical Identifiers
| 77-93-0 | |
| 276.285 | |
| DOOTYTYQINUNNV-UHFFFAOYSA-N | |
| 6506 | |
| CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O |
| C12H20O7 | |
| MFCD00009201 | |
| triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
| triethyl 2-hydroxypropane-1,2,3-tricarboxylate |
Safety and Handling
EINECSNumber : 201-070-7
RTECSNumber : GE8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only