missing translation for 'onlineSavingsMsg'
Learn More

Triethyl citrate, 99%

Catalog No. p-7045969
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1263914 25 g
AAL1263930 250 g
AAL126390B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1263914 Supplier Thermo Scientific Chemicals Supplier No. L1263914
Only null left

CAS: 77-93-0 | C12H20O7 | 276.285 g/mol

Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.285
MDL Number MFCD00009201
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
Synonym triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
PubChem CID 6506
IUPAC Name triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O

Specifications

Melting Point -55°C
Density 1.137
Boiling Point 126°C to 127°C (1 mmHg)
Flash Point 176°C (348°F)
Refractive Index 1.442
Quantity 25 g
Beilstein 1801199
Merck Index 14,2326
Solubility Information Miscible with water,alcohol and diethyl ether.
Formula Weight 276.29
Percent Purity 99%
Chemical Name or Material Triethyl citrate
Show More Show Less
EINECSNumber 201-070-7
RTECSNumber GE8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only