missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl 2-Phosphonopropionate 98.0+%, TCI America™
Supplier: TCI America T213525G
Specifications
| Triethyl 2-Phosphonopropionate | |
| Colorless | |
| C9H19O5P | |
| 25 g | |
| BVSRWCMAJISCTD-UHFFFAOYSA-N | |
| ethyl 2-diethoxyphosphorylpropanoate | |
| 107155 | |
| ≥98.0% (GC) |
| 3699-66-9 | |
| 144°C | |
| MFCD00009159 | |
| triethyl 2-phosphonopropionate, ethyl 2-diethoxyphosphoryl propanoate, ethyl-2-diethylphosphono propanoate, diethyl 1-carbethoxy ethylphosphonate, propanoic acid, 2-diethoxyphosphinyl-, ethyl ester, diethyl ethoxycarbonylethyl-phosphonate, diethyl 1-ethoxycarbonyl ethanephosphonate, 2-diethoxyphosphinyl propanoic acid ethyl ester, diethyl 1-ethoxycarbonyl ethanephosphonate∼2-phosphonopropionic acid triethyl ester, triethyl2-phosphonopropionate | |
| CCOC(=O)C(C)P(=O)(OCC)OCC | |
| 238.22 | |
| 238.22 | |
| Liquid |
Chemical Identifiers
| 3699-66-9 | |
| 238.22 | |
| BVSRWCMAJISCTD-UHFFFAOYSA-N | |
| ethyl 2-diethoxyphosphorylpropanoate |
| C9H19O5P | |
| MFCD00009159 | |
| 107155 | |
| CCOC(=O)C(C)P(=O)(OCC)OCC |
Safety and Handling
TSCA : No