missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-Phosphonopropionate 98.0+%, TCI America™

Catalog No. p-7137097
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21355G 5 g
T213525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21355G Supplier TCI America Supplier No. T21355G

Chemical Identifiers

CAS 3699-66-9
Molecular Formula C9H19O5P
Molecular Weight (g/mol) 238.22
MDL Number MFCD00009159
InChI Key BVSRWCMAJISCTD-UHFFFAOYSA-N
PubChem CID 107155
IUPAC Name ethyl 2-diethoxyphosphorylpropanoate
SMILES CCOC(=O)C(C)P(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 144°C
Quantity 5 g
Synonym triethyl 2-phosphonopropionate, ethyl 2-diethoxyphosphoryl propanoate, ethyl-2-diethylphosphono propanoate, diethyl 1-carbethoxy ethylphosphonate, propanoic acid, 2-diethoxyphosphinyl-, ethyl ester, diethyl ethoxycarbonylethyl-phosphonate, diethyl 1-ethoxycarbonyl ethanephosphonate, 2-diethoxyphosphinyl propanoic acid ethyl ester, diethyl 1-ethoxycarbonyl ethanephosphonate∼2-phosphonopropionic acid triethyl ester, triethyl2-phosphonopropionate
Formula Weight 238.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl 2-Phosphonopropionate