missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 1,3,5-benzenetricarboxylate, 97%

Catalog No. AAB2339214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2339214 25 g
AAB2339206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2339214 Supplier Thermo Scientific Chemicals Supplier No. B2339214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4105-92-4 | C15H18O6 | 294.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4105-92-4
Molecular Formula C15H18O6
Molecular Weight (g/mol) 294.303
MDL Number MFCD00236389
InChI Key KXGOWZRHSOJOLF-UHFFFAOYSA-N
Synonym triethyl trimesate, 1,3,5-benzenetricarboxylic acid, triethyl ester, trimesic acid triethyl ester, triethyl 1,3,5-benzenetricarboxylate, ethyl 3,5-bis ethoxycarbonyl benzoate, 1,3,5-triethyl benzene-1,3,5-tricarboxylate, acmc-20a6fr, bzta02, 1,3,5-tri ethoxycarbonyl benzene, triethyl 1,5-benzenetricarboxylate
PubChem CID 226764
IUPAC Name triethyl benzene-1,3,5-tricarboxylate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC

Specifications

Melting Point 132°C to 135°C
Quantity 25 g
Formula Weight 294.31
Percent Purity 97%
Chemical Name or Material Triethyl 1,3,5-benzenetricarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only