CAS RN 4105-92-4
IUPAC Name:
1,3,5-triethyl benzene-1,3,5-tricarboxylate
Synonyms:
1,3,5-triethyl benzene-1,3,5-tricarboxylate
Molecular Weight (g/mol):
294.3
Molecular Formula:
C15H18O6
InChi Key:
KXGOWZRHSOJOLF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC
1
–
1
of
1
results
1
Promotions Available
Triethyl 1,3,5-benzenetricarboxylate, 97%
Triethyl 1,3,5-benzenetricarboxylate, 97%
CAS: 4105-92-4 Molecular Formula: C15H18O6 Molecular Weight (g/mol): 294.303 MDL Number: MFCD00236389 InChI Key: KXGOWZRHSOJOLF-UHFFFAOYSA-N Synonym: triethyl trimesate,1,3,5-benzenetricarboxylic acid, triethyl ester,trimesic acid triethyl ester,triethyl 1,3,5-benzenetricarboxylate,ethyl 3,5-bis ethoxycarbonyl benzoate,1,3,5-triethyl benzene-1,3,5-tricarboxylate,acmc-20a6fr,bzta02,1,3,5-tri ethoxycarbonyl benzene,triethyl 1,5-benzenetricarboxylate PubChem CID: 226764 IUPAC Name: triethyl benzene-1,3,5-tricarboxylate SMILES: CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC