missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 1,1,2-Ethanetricarboxylate 96.0+%, TCI America™

Catalog No. p-7137215
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24315G 5 g
T243125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24315G Supplier TCI America Supplier No. T24315G

Chemical Identifiers

CAS 7459-46-3
Molecular Formula C11H18O6
Molecular Weight (g/mol) 246.259
MDL Number MFCD00009154
InChI Key TVWZLLYAJDSSCJ-UHFFFAOYSA-N
Synonym triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
PubChem CID 81961
IUPAC Name triethyl ethane-1,1,2-tricarboxylate
SMILES CCOC(=O)CC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 165°C
Quantity 5 g
Formula Weight 246.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl 1,1,2-Ethanetricarboxylate