accessibility menu, dialog, popup

UserName

H3COOOOCH3OOCH3

CAS RN 7459-46-3

IUPAC Name: 1,1,2-triethyl ethane-1,1,2-tricarboxylate
Synonyms: 1,1,2-triethyl ethane-1,1,2-tricarboxylate
Molecular Weight (g/mol): 246.26
Molecular Formula: C11H18O6
InChi Key: TVWZLLYAJDSSCJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)