missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyvinylsilane 98.0+%, TCI America™

Catalog No. p-7137899
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V004425ML 25 mL
V0044100ML 100 mL
V0044500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V004425ML Supplier TCI America Supplier No. V004425ML
Only null left

Chemical Identifiers

CAS 78-08-0
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00009063
InChI Key FWDBOZPQNFPOLF-UHFFFAOYSA-N
Synonym vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
PubChem CID 6516
IUPAC Name ethenyltriethoxysilane
SMILES CCO[Si](OCC)(OCC)C=C

Specifications

Color Colorless
Boiling Point 161°C
Quantity 25 mL
UN Number 1993
Formula Weight 190.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxyvinylsilane
EINECSNumber (2)-2066
RTECSNumber VV6700000
TSCA Yes