accessibility menu, dialog, popup

UserName

CAS RN 78-08-0

IUPAC Name: ethenyltriethoxysilane
Synonyms: triethoxyvinylsilane
Molecular Weight (g/mol): 190.31
Molecular Formula: C8H18O3Si
InChi Key: FWDBOZPQNFPOLF-UHFFFAOYSA-N
SMILES: CCO[Si](OCC)(OCC)C=C
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (3)
  • (5)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)