missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyfluorosilane 93.0+%, TCI America™

Catalog No. T16965ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16965ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16965ML Supplier TCI America Supplier No. T16965ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 358-60-1
Molecular Formula C6H15FO3Si
Molecular Weight (g/mol) 182.266
MDL Number MFCD00236719
InChI Key XVYIJOWQJOQFBG-UHFFFAOYSA-N
Synonym fluorotriethoxysilane, triethoxy fluoro silane, silane, triethoxyfluoro, triethoxyfluorosilicon, silicon fluoride triethoxide, fluoro triethoxy silan, triethoxy fluoranyl silane, acmc-1cqk0
PubChem CID 2776872
IUPAC Name triethoxy(fluoro)silane
SMILES CCO[Si](OCC)(OCC)F

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 mL
UN Number 2924
Formula Weight 182.27
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxyfluorosilane