missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy(3-glycidyloxypropyl)silane 96.0+%, TCI America™

Catalog No. T26755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26755G 5 g
T267525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26755G Supplier TCI America Supplier No. T26755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2602-34-8
Molecular Formula C12H26O5Si
Molecular Weight (g/mol) 278.42
MDL Number MFCD01311705
InChI Key JXUKBNICSRJFAP-UHFFFAOYNA-N
Synonym Glycidyl 3-Triethoxysilylpropyl Ether
PubChem CID 102856
IUPAC Name triethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
SMILES CCO[Si](CCCOCC1CO1)(OCC)OCC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 5 g
Formula Weight 278.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy(3-glycidyloxypropyl)silane
EINECSNumber (2)-2071
TSCA Yes