CAS RN 2602-34-8
IUPAC Name:
triethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
Synonyms:
triethoxy[3-(oxiran-2-ylmethoxy)propyl]silane
Molecular Weight (g/mol):
278.42
Molecular Formula:
C12H26O5Si
InChi Key:
JXUKBNICSRJFAP-UHFFFAOYNA-N
SMILES:
CCO[Si](CCCOCC1CO1)(OCC)OCC
1
–
1
of
1
results
1
Triethoxy(3-glycidyloxypropyl)silane 96.0+%, TCI America™
CAS: 2602-34-8 Molecular Formula: C12H26O5Si Molecular Weight (g/mol): 278.42 MDL Number: MFCD01311705 InChI Key: JXUKBNICSRJFAP-UHFFFAOYNA-N Synonym: Glycidyl 3-Triethoxysilylpropyl Ether PubChem CID: 102856 IUPAC Name: triethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane SMILES: CCO[Si](CCCOCC1CO1)(OCC)OCC