missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane 98.0+%, TCI America™

Catalog No. t35855ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35855ML 5 mL
T358525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35855ML Supplier TCI America Supplier No. T35855ML
Only null left

Chemical Identifiers

CAS 10217-34-2
Molecular Formula C14H28O4Si
Molecular Weight (g/mol) 288.46
MDL Number MFCD00069159
InChI Key UDUKMRHNZZLJRB-UHFFFAOYNA-N
Synonym 2-(3,4-Epoxycyclohexyl)ethyltriethoxysilane
PubChem CID 154915
IUPAC Name triethoxy(2-{7-oxabicyclo[4.1.0]heptan-3-yl}ethyl)silane
SMILES CCO[Si](CCC1CCC2OC2C1)(OCC)OCC

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 mL
Formula Weight 288.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
EINECSNumber (3)-2647
TSCA Yes