accessibility menu, dialog, popup

UserName

CAS RN 10217-34-2

IUPAC Name: triethoxy(2-{7-oxabicyclo[4.1.0]heptan-3-yl}ethyl)silane
Synonyms: triethoxy(2-{7-oxabicyclo[4.1.0]heptan-3-yl}ethyl)silane
Molecular Weight (g/mol): 288.46
Molecular Formula: C14H28O4Si
InChi Key: UDUKMRHNZZLJRB-UHFFFAOYNA-N
SMILES: CCO[Si](CCC1CCC2OC2C1)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results