missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane 97.0+%, TCI America™
Supplier: TCI America T177025G
Specifications
| Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane | |
| Yellow | |
| C14H19F13O3Si | |
| 25 g | |
| AVYKQOAMZCAHRG-UHFFFAOYSA-N | |
| triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane | |
| 103991 | |
| ≥97.0% (GC) |
| 51851-37-7 | |
| 86°C | |
| MFCD00042333 | |
| 1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane | |
| CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC | |
| 510.367 | |
| 510.37 | |
| Liquid |
Chemical Identifiers
| 51851-37-7 | |
| 510.367 | |
| AVYKQOAMZCAHRG-UHFFFAOYSA-N | |
| 103991 | |
| CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC |
| C14H19F13O3Si | |
| MFCD00042333 | |
| 1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane | |
| triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
Safety and Handling
TSCA : Yes