missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane 97.0+%, TCI America™

Catalog No. p-7136951
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17705G 5 g
T177025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17705G Supplier TCI America Supplier No. T17705G
Only null left

Chemical Identifiers

CAS 51851-37-7
Molecular Formula C14H19F13O3Si
Molecular Weight (g/mol) 510.367
MDL Number MFCD00042333
InChI Key AVYKQOAMZCAHRG-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane
PubChem CID 103991
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 510.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane