missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97.0+%, TCI America™

Catalog No. p-7137455
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T28605G 5 g
T286025G 25 g
2 options

Catalog No. T28605G

Supplier: TCI America T28605G

Only null left

Chemical Identifiers

CAS 102390-98-7
Molecular Formula C12H19F9O3Si
Molecular Weight (g/mol) 410.352
InChI Key NYIKUOULKCEZDO-UHFFFAOYSA-N
Synonym 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane
PubChem CID 22600765
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
SMILES CCO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 410.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane