accessibility menu, dialog, popup

UserName

CAS RN 102390-98-7

IUPAC Name: triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
Synonyms: triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
Molecular Weight (g/mol): 410.35
Molecular Formula: C12H19F9O3Si
InChi Key: NYIKUOULKCEZDO-UHFFFAOYSA-N
SMILES: CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results