missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012321
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012321 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012321 Supplier Merck Emd Millipore Supplier No. 509261ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 102-71-6
Ignition Point 600°F
Vapor Pressure 0.01 mmHg (20°C)
Vapor Density 5.14 (vs. air)
Density 1.124 g/mL (at 25°C)
Flash Point 179°C
Physical Form Neat
Linear Formula (HOCH2 CH2)3 N
MDL Number MFCD00002855
Melting Point 17.9°C to 21°C
Boiling Point 190°C to 193°C (5 mmHg)
Synonym 2,2′,2′′-Nitrilotriethanol; Tris(2-hydroxyethyl)amine
Solubility Soluble
Type Analytical Standard
Quantity 1 mL
Formula Weight 149.19
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less