missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 50% v/v, Reagents

Catalog No. 01333413
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333413 1 L
01333412 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333413 Supplier Labchem Inc Supplier No. CT1530001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triethanolamine, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 102-71-6 , 7732-18-5
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym 2,2,2-Nitrilotriethanol, Trolamine
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Color Colorless to Pale Yellow
Boiling Point 335°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.073
Percent Purity 50%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethanolamine, 50% v/v