missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trielaidin 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 42°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 885.45 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trielaidin |
Safety and Handling
| EINECSNumber | (2)-0669 |
| TSCA | No |
| Recommended Storage | Freezer |