missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tridecylbenzene 99.0+%, TCI America™
Supplier: TCI America T04195ML
Specifications
| Tridecylbenzene | |
| 10°C | |
| 346°C | |
| MFCD00008980 | |
| 1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs. | |
| CCCCCCCCCCCCCC1=CC=CC=C1 | |
| 260.465 | |
| 260.47 | |
| Liquid |
| 123-02-4 | |
| Colorless | |
| C19H32 | |
| 5 mL | |
| MCVUKOYZUCWLQQ-UHFFFAOYSA-N | |
| tridecylbenzene | |
| 31238 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 123-02-4 | |
| 260.465 | |
| MCVUKOYZUCWLQQ-UHFFFAOYSA-N | |
| 31238 | |
| CCCCCCCCCCCCCC1=CC=CC=C1 |
| C19H32 | |
| MFCD00008980 | |
| 1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs. | |
| tridecylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes