accessibility menu, dialog, popup

UserName

CAS RN 123-02-4

H3C

CAS RN 123-02-4

IUPAC Name: tridecylbenzene
Synonyms: 1-phenyltridecane
Molecular Weight (g/mol): 260.47
Molecular Formula: C19H32
InChi Key: MCVUKOYZUCWLQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results