missing translation for 'onlineSavingsMsg'
Learn More

Tridecyl Borate 98.0+%, TCI America™

Catalog No. B20965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20965G Supplier TCI America Supplier No. B20965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20236-81-1
Molecular Formula C30H63BO3
Molecular Weight (g/mol) 482.641
MDL Number MFCD00026531
InChI Key WAXLMVCEFHKADZ-UHFFFAOYSA-N
Synonym Boric Acid Tridecyl Ester
PubChem CID 88422
IUPAC Name tris-decyl borate
SMILES B(OCCCCCCCCCC)(OCCCCCCCCCC)OCCCCCCCCCC

Specifications

Color Yellow
Boiling Point 258°C
Quantity 5 g
Formula Weight 482.64
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tridecyl Borate