accessibility menu, dialog, popup

UserName

CAS RN 20236-81-1

CH3OBOH3COCH3

CAS RN 20236-81-1

IUPAC Name: tris(decyl) borate
Synonyms: tris(decyl) borate
Molecular Weight (g/mol): 482.64
Molecular Formula: C30H63BO3
InChi Key: WAXLMVCEFHKADZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOB(OCCCCCCCCCC)OCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results