missing translation for 'onlineSavingsMsg'
Learn More

Tridecanophenone 96.0+%, TCI America™

Catalog No. T108710G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T108710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T108710G Supplier TCI America Supplier No. T108710G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6005-99-8
Molecular Formula C19H30O
Molecular Weight (g/mol) 274.45
MDL Number MFCD00059219
InChI Key ZZNDQCACFUJAKJ-UHFFFAOYSA-N
Synonym Dodecyl Phenyl Ketone
PubChem CID 577616
IUPAC Name 1-phenyltridecan-1-one
SMILES CCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 10 g
Formula Weight 274.45
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tridecanophenone